ENA946840019 Enamine

1-(4-aminophenyl)-1,3-dimethylurea

  • MDL number MFCD24262165
  • Cas Number : 1556187-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379065
Availability details for
Quantity
  • Availability
  • Add to Cart