ENA946840025 Enamine

3-(1-methyl-1H-pyrazol-4-yl)-8-azabicyclo[3.2.1]octane

  • MDL number MFCD24506198
  • Cas Number : 1691745-69-3
  • Molecular Weight : 191.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379077
Availability details for
Quantity
  • Availability
  • Add to Cart