ENA946840037 Enamine

ethyl 5-(cyclopropylamino)-1,3,4-oxadiazole-2-carboxylate

  • MDL number MFCD30734059
  • Cas Number : 2094327-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379163
Availability details for
Quantity
  • Availability
  • Add to Cart