ENA946840055 Enamine

potassium [(3-bromophenyl)methyl]trifluoroboranuide

  • MDL number MFCD18861713
  • Molecular Weight : 276.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379223
Availability details for
Quantity
  • Availability
  • Add to Cart