ENA946840062 Enamine

2-(2-aminoethyl)-1lambda6-thiolane-1,1-dione hydrochloride

  • MDL number MFCD30002755
  • Cas Number : 2094938-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379472
Availability details for
Quantity
  • Availability
  • Add to Cart