ENA946840079 Enamine

3-(3-bromo-2-methylphenyl)-8-fluoro-1-methyl-1,2,3,4-tetrahydroquinazoline-2,4-dione

  • MDL number MFCD30734074
  • Cas Number : 1643136-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379497
Availability details for
Quantity
  • Availability
  • Add to Cart