ENA946840084 Enamine

2-(2H-1,2,3-triazol-2-yl)-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD28967297
  • Cas Number : 1435479-65-4
  • Molecular Weight : 257.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379512

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379512
Availability details for
Quantity
  • Availability
  • Add to Cart