ENA946840085 Enamine

2-sulfanylpyridine-3-carbothioamide

  • MDL number MFCD26674633
  • Cas Number : 1529146-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379514
Availability details for
Quantity
  • Availability
  • Add to Cart