ENA946840092 Enamine

2-[methyl(phenyl)amino]-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD16870524
  • Cas Number : 1339523-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379535
Availability details for
Quantity
  • Availability
  • Add to Cart