ENA946840099 Enamine

4-bromo-2-(chloromethyl)-1-(3,4-dichlorophenoxy)benzene

  • MDL number MFCD30734081
  • Cas Number : 1929328-19-7
  • Molecular Weight : 366.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379595
Availability details for
Quantity
  • Availability
  • Add to Cart