ENA946840120 Enamine

6-fluoro-2-hydroxyquinoline-3-carboxylic acid

  • MDL number MFCD18445722
  • Cas Number : 938378-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379658
Availability details for
Quantity
  • Availability
  • Add to Cart