ENA946840123 Enamine

ethyl 4-chlorobenzene-1-sulfinate

  • MDL number MFCD08460264
  • Cas Number : 13165-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379662

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P272,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P333+P313,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-379662
pictograms GHS05,GHS07
hstatements H302,H314,H317
Availability details for
Quantity
  • Availability
  • Add to Cart