ENA946840127 Enamine

2-[5-(2-chlorophenyl)-1,3,4-oxadiazol-2-yl]benzoic acid

  • MDL number MFCD30585110
  • Cas Number : 56894-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379671
Availability details for
Quantity
  • Availability
  • Add to Cart