ENA946840141 Enamine

5-amino-3-methyl-2,3-dihydro-1,3-benzothiazol-2-one

  • MDL number MFCD20703803
  • Cas Number : 1379344-15-6
  • Molecular Weight : 180.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-380822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-380822
Availability details for
Quantity
  • Availability
  • Add to Cart