ENA946840156 Enamine

5-phenyl-2-(tributylstannyl)-1,3-oxazole

  • MDL number MFCD13181862
  • Cas Number : 1019654-81-9
  • Molecular Weight : 434.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-380895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-380895
Availability details for
Quantity
  • Availability
  • Add to Cart