ENA946840161 Enamine

(methylsulfanyl)ethene

  • MDL number MFCD00085292
  • Cas Number : 1822-74-8
  • Molecular Weight : 74.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-380910

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P280,P303+P361+P353,P305+P351+P338,P310,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-380910
pictograms GHS02,GHS05
hstatements H225,H318
Availability details for
Quantity
  • Availability
  • Add to Cart