ENA946840175 Enamine

4-bromo-5-chloro-2-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD12026191
  • Cas Number : 1208076-84-9
  • Molecular Weight : 303.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-380959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-380959
Availability details for
Quantity
  • Availability
  • Add to Cart