ENA946840184 Enamine

2-{[(5-bromo-1,3-thiazol-2-yl)carbamoyl]methoxy}acetic acid

  • MDL number MFCD14620839
  • Cas Number : 1248470-13-4
  • Molecular Weight : 295.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-381003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-381003
Availability details for
Quantity
  • Availability
  • Add to Cart