ENA946840210 Enamine

1-(pyrimidin-2-yl)cyclobutan-1-amine

  • MDL number MFCD28645540
  • Cas Number : 1782058-56-3
  • Molecular Weight : 149.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-381042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-381042
Availability details for
Quantity
  • Availability
  • Add to Cart