ENA946840217 Enamine

4-[(2-fluorophenyl)methoxy]phenol

  • MDL number MFCD11181764
  • Cas Number : 1094247-88-7
  • Molecular Weight : 218.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-381062

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-381062
Availability details for
Quantity
  • Availability
  • Add to Cart