ENA946840231 Enamine

tert-butyl 2-(4-fluorophenyl)-2-oxoacetate

  • MDL number MFCD30733991
  • Cas Number : 1070237-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378043
Availability details for
Quantity
  • Availability
  • Add to Cart