ENA946840233 Enamine

3-methyl-1,3-oxazolidine

  • MDL number MFCD01938319
  • Cas Number : 27970-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-381121

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P280,P303+P361+P353,P305+P351+P338,P310,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-381121
pictograms GHS02,GHS05
hstatements H225,H318
Availability details for
Quantity
  • Availability
  • Add to Cart