ENA946840251 Enamine

3-bromo-5-(oxiran-2-yl)-1,2-oxazole

  • MDL number MFCD30733994
  • Cas Number : 76596-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378196
Availability details for
Quantity
  • Availability
  • Add to Cart