ENA946840253 Enamine

(2,3-difluorophenyl)(2-fluoro-4-methoxyphenyl)methanol

  • MDL number MFCD17372625
  • Cas Number : 1271539-73-1
  • Molecular Weight : 268.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-381187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-381187
Availability details for
Quantity
  • Availability
  • Add to Cart