ENA946840263 Enamine

2-{2-[(tert-butoxy)carbonyl]-2-azaspiro[3.3]heptan-5-yl}acetic acid

  • MDL number MFCD27996147
  • Cas Number : 1341036-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378245
Availability details for
Quantity
  • Availability
  • Add to Cart