ENA946840266 Enamine

tert-butyl N-[(1R,3R,4S)-3,4-dihydroxycyclopentyl]carbamate

  • MDL number MFCD29921625
  • Cas Number : 1853339-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378250
Availability details for
Quantity
  • Availability
  • Add to Cart