ENA946840270 Enamine

4-hydroxy-1-oxo-1,2-dihydroisoquinoline-3-carboxamide

  • MDL number MFCD24563053
  • Cas Number : 99275-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383233
Availability details for
Quantity
  • Availability
  • Add to Cart