ENA946840277 Enamine

4-bromopent-1-ene

  • MDL number MFCD20621242
  • Cas Number : 31950-56-8
  • Molecular Weight : 149.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383267

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P271,P280,P285,P303+P361+P353,P304+P340,P304+P341,P305+P351+P338,P312,P337+P313,P342+P311,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-383267
pictograms GHS02,GHS08
hstatements H225,H319,H334,H335
Availability details for
Quantity
  • Availability
  • Add to Cart