ENA946840278 Enamine

4-methyl-3-(morpholine-4-sulfonyl)-N-[4-(pyridin-2-yl)-1,3-thiazol-2-yl]benzamide

  • MDL number MFCD07321219
  • Cas Number : 848373-17-1
  • Molecular Weight : 444.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378278
Availability details for
Quantity
  • Availability
  • Add to Cart