ENA946840280 Enamine

methyl 5-methyl-4,5,6,7-tetrahydro-1H-indazole-4-carboxylate

  • MDL number MFCD30733996
  • Cas Number : 2094408-06-5
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378320

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378320
Availability details for
Quantity
  • Availability
  • Add to Cart