ENA946840282 Enamine

4-methyl-2-(tributylstannyl)pyrimidine

  • MDL number MFCD29111158
  • Cas Number : 1447763-68-9
  • Molecular Weight : 383.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383277

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P273,P280,P301+P310,P302+P352,P304+P340,P305+P351+P338,P312,P314,P330,P332+P313,P337+P313,P362,P363,P391,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-383277
pictograms GHS06,GHS08,GHS09
hstatements H301,H312,H315,H319,H335,H372,H400,H410
Availability details for
Quantity
  • Availability
  • Add to Cart