ENA946840287 Enamine

3-benzyl-1,2,4-oxadiazole-5-thiol

  • MDL number MFCD03069569
  • Cas Number : 494780-00-6
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383289
Availability details for
Quantity
  • Availability
  • Add to Cart