ENA946840311 Enamine

4-(dimethylamino)pyridine-2-carbonitrile

  • MDL number MFCD17252590
  • Cas Number : 896139-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383763
Availability details for
Quantity
  • Availability
  • Add to Cart