ENA946840312 Enamine

tert-butyl 2,7-diazaspiro[4.6]undecane-2-carboxylate

  • MDL number MFCD12406562
  • Cas Number : 1205750-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383766
Availability details for
Quantity
  • Availability
  • Add to Cart