ENA946840316 Enamine

2-benzyl-2-methyl-1,3-thiazolidine

  • MDL number MFCD01681132
  • Cas Number : 773-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383771
Availability details for
Quantity
  • Availability
  • Add to Cart