ENA946840318 Enamine

1-cyclopropylcyclobutane-1-carboxylic acid

  • MDL number MFCD28148315
  • Cas Number : 1498554-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383773
Availability details for
Quantity
  • Availability
  • Add to Cart