ENA946840324 Enamine

(1r,3r)-3-fluoro-1-phenylcyclobutane-1-carboxylic acid

  • MDL number MFCD30734166
  • Cas Number : 1932284-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383784
Availability details for
Quantity
  • Availability
  • Add to Cart