ENA946840326 Enamine

(2E)-N-phenylbut-2-enamide

  • MDL number MFCD23713287
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383792
Availability details for
Quantity
  • Availability
  • Add to Cart