ENA946840333 Enamine

5-(benzyloxy)-2-(propan-2-yl)pyrimidine-4-carboxylic acid

  • MDL number MFCD28650373
  • Cas Number : 1455280-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383822
Availability details for
Quantity
  • Availability
  • Add to Cart