ENA946840337 Enamine

3-tert-butyl-4-propoxybenzene-1-sulfonamide

  • MDL number MFCD11213172
  • Cas Number : 1094537-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383831
Availability details for
Quantity
  • Availability
  • Add to Cart