ENA946840378 Enamine

5-cyclohexyl-1,2,4-thiadiazol-3-amine

  • MDL number MFCD20348081
  • Cas Number : 1344332-83-7
  • Molecular Weight : 183.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378679
Availability details for
Quantity
  • Availability
  • Add to Cart