ENA946840387 Enamine

3-(hydroxymethyl)-4-methoxybenzene-1-sulfonamide

  • MDL number MFCD21298889
  • Cas Number : 900165-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383987
Availability details for
Quantity
  • Availability
  • Add to Cart