ENA946840390 Enamine

(1S,2S)-2-methylcyclopentan-1-ol

  • MDL number MFCD20760177
  • Cas Number : 39947-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 47
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383998

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P280,P303+P361+P353,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-383998
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart