ENA946840392 Enamine

1-bromo-4-nitroisoquinoline

  • MDL number MFCD00834981
  • Cas Number : 55404-30-3
  • Molecular Weight : 253.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-125
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378712
Availability details for
Quantity
  • Availability
  • Add to Cart