ENA946840402 Enamine

2-fluoro-1,2,3,4-tetrahydronaphthalen-1-ol

  • MDL number MFCD24392149
  • Cas Number : 1824069-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384043
Availability details for
Quantity
  • Availability
  • Add to Cart