ENA946840444 Enamine

2-amino-1-(1,3-thiazol-5-yl)ethan-1-ol dihydrochloride

  • MDL number MFCD28895295
  • Cas Number : 2094879-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384159
Availability details for
Quantity
  • Availability
  • Add to Cart