ENA946840451 Enamine

N-(2-methyl-2H-1,2,3,4-tetrazol-5-yl)-2-(methylamino)benzene-1-sulfonamide

  • MDL number MFCD25142352
  • Cas Number : 1538917-85-9
  • Molecular Weight : 268.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384171
Availability details for
Quantity
  • Availability
  • Add to Cart