ENA946840464 Enamine

5-(methoxycarbonyl)-3-methylfuran-2-carboxylic acid

  • MDL number MFCD30734211
  • Cas Number : 2090165-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384199
Availability details for
Quantity
  • Availability
  • Add to Cart