ENA946840475 Enamine

3-(azetidin-3-yl)-5-methyl-1H-1,2,4-triazole dihydrochloride

  • MDL number MFCD28894690
  • Cas Number : 2095409-71-3
  • Molecular Weight : 211.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384241
Availability details for
Quantity
  • Availability
  • Add to Cart