ENA946840486 Enamine

(1S)-4-fluoro-2,3-dihydro-1H-inden-1-ol

  • MDL number MFCD15529824
  • Cas Number : 1270289-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384277
Availability details for
Quantity
  • Availability
  • Add to Cart